Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CCKBR
Entrez Gene ID
100328939
Gene Name
cholecystokinin B receptor
Gene Aliases
CCK-B, CCK-BR, CCK2-R
Chromosome Location
Chr.1: 147270738 - 147284108 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001171123.2 NP_001164594.2 2 - 3 803 57

View Full Product Details