Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SERPINB13
Entrez Gene ID
100328644
Gene Name
serpin family B member 13
Gene Aliases
-
Chromosome Location
Chr.9: 103823966 - 103846703 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001171283.1 NP_001164754.1 4 - 5 470 88
XM_008261382.2 XP_008259604.1 5 - 6 554 88
XM_008261383.2 XP_008259605.1 5 - 6 617 88
XM_008261384.2 XP_008259606.1 4 - 5 497 88

View Full Product Details