Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SERPINB7
Entrez Gene ID
100328647
Gene Name
serpin family B member 7
Gene Aliases
-
Chromosome Location
Chr.9: 103966796 - 104056172 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001171286.1 NP_001164757.1 4 - 5 455 119
XM_008261387.2 XP_008259609.1 5 - 6 523 119
XM_008261388.2 XP_008259610.1 5 - 6 562 119
XM_017343654.1 XP_017199143.1 5 - 6 600 119

View Full Product Details