Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ABCD1
Entrez Gene ID
100328741
Gene Name
ATP binding cassette subfamily D member 1
Gene Aliases
-
Chromosome Location
Chr.Unknown: 677038 - 693729 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001171362.1 NP_001164833.1 - 1994 65

View Full Product Details