Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GAB3
Entrez Gene ID
100328766
Gene Name
GRB2 associated binding protein 3
Gene Aliases
-
Chromosome Location
Chr.Unknown: 954811 - 1037426 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001171383.1 NP_001164854.1 - 1433 99
XM_008249459.2 XP_008247681.1 - 1467 99

View Full Product Details