Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HOXA2
Entrez Gene ID
100328783
Gene Name
homeobox A2
Gene Aliases
-
Chromosome Location
Chr.10: 11820487 - 11822278 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001171398.1 NP_001164869.1 1 - 2 392 97

View Full Product Details