Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CYP2G1
Entrez Gene ID
100359338
Gene Name
cytochrome P450 2G1
Gene Aliases
CYPIIG1
Chromosome Location
Chr.Unknown: 884179 - 894850 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001195764.1 NP_001182693.1 - 972 85

View Full Product Details