Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SELENOI
Entrez Gene ID
100354509
Gene Name
selenoprotein I
Gene Aliases
EPT1, SELI
Chromosome Location
Chr.2: 159531244 - 159569255 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001199841.1 NP_001186770.1 2 - 3 265 68

View Full Product Details