Gene Transcripts

Gene Symbol
Gid8
Entrez Gene ID
296466
Gene Name
GID complex subunit 8
Gene Aliases
Bwk1, TWA1
Chromosome Location
Chr. 3: 176217128 - 176223645 on Build Rnor_6.0
UniGene ID
Rn.28608
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198743.1 NP_942038.1 5 606
GenBank mRNA AY156921.1 - 5 606

View Full Product Details