Gene Transcripts

Gene Symbol
Fam81b
Entrez Gene ID
309925
Gene Name
family with sequence similarity 81, member B
Gene Aliases
RGD1561830
Chromosome Location
Chr. 2: 3004639 - 3124537 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008760605.1 XP_008758827.1 5, 6 625

View Full Product Details