Gene Transcripts

Gene Symbol
Pop1
Entrez Gene ID
315045
Gene Name
processing of precursor 1, ribonuclease P/MRP subunit (S. cerevisiae)
Gene Aliases
Chromosome Location
Chr. 7: 73270411 - 73298209 on Build Rnor_6.0
UniGene ID
Rn.22746
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001130550.1 NP_001124022.1 11 1589
XM_006241543.2 XP_006241605.1 11 1633
GenBank mRNA FQ223904.1 - 11 1608

View Full Product Details