Gene Transcripts

Gene Symbol
Robo3
Entrez Gene ID
315564
Gene Name
roundabout, axon guidance receptor, homolog 3 (Drosophila)
Gene Aliases
Chromosome Location
Chr. 8: 39892792 - 39910596 on Build Rnor_6.0
UniGene ID
Rn.45295
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108135.1 NP_001101605.1 8 1581
XM_008766057.1 XP_008764279.1 9 1706

View Full Product Details