Gene Transcripts

Gene Symbol
Elmod1
Entrez Gene ID
315670
Gene Name
ELMO/CED-12 domain containing 1
Gene Aliases
RGD1564186
Chromosome Location
Chr. 8: 58486078 - 58542888 on Build Rnor_6.0
UniGene ID
Rn.13823
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001191579.1 NP_001178508.1 6 598
XM_006243083.2 XP_006243145.1 6 642
XM_008766269.1 XP_008764491.1 6 680

View Full Product Details