Gene Transcripts

Gene Symbol
Sphkap
Entrez Gene ID
316561
Gene Name
SPHK1 interactor, AKAP domain containing
Gene Aliases
RGD1311951, Skip
Chromosome Location
Chr. 9: 89050405 - 89195445 on Build Rnor_6.0
UniGene ID
Rn.15453
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001127492.1 NP_001120964.1 9 3309
XM_006245230.2 XP_006245292.1 8 3538
XM_006245231.2 XP_006245293.1 8 3939
XM_006245233.2 XP_006245295.1 7 3458
XM_006245234.2 XP_006245296.1 8 3537
XM_008767251.1 XP_008765473.1 7 3860

View Full Product Details