Gene Transcripts

Gene Symbol
R3hcc1
Entrez Gene ID
361064
Gene Name
R3H domain and coiled-coil containing 1
Gene Aliases
RGD1311790
Chromosome Location
Chr. 15: 51368938 - 51385087 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006252286.2 XP_006252348.1 7, 8 1664
XM_006252289.1 XP_006252351.1 7, 8 1453

View Full Product Details