Gene Transcripts

Gene Symbol
Tango6
Entrez Gene ID
307816
Gene Name
transport and golgi organization 6 homolog (Drosophila)
Gene Aliases
RGD1559841
Chromosome Location
Chr. 19: 38846204 - 39050217 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006255568.2 XP_006255630.1 10 1776
XM_008772562.1 XP_008770784.1 11 1953
XM_226428.8 XP_226428.6 11 1953

View Full Product Details