Gene Transcripts

Gene Symbol
Fam73a
Entrez Gene ID
362058
Gene Name
family with sequence similarity 73, member A
Gene Aliases
RGD1559958
Chromosome Location
Chr. 2: 257490729 - 257546850 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008761559.1 XP_008759781.1 2 176

View Full Product Details