Gene Transcripts

Gene Symbol
Slc9b1
Entrez Gene ID
365946
Gene Name
solute carrier family 9, subfamily B (NHA1, cation proton antiporter 1), member 1
Gene Aliases
Nhedc1, Nhedc2, RGD1305087
Chromosome Location
Chr. 2: 240527130 - 240576179 on Build Rnor_6.0
UniGene ID
Rn.133113
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001173973.1 NP_001167444.1 8 1294
XM_008761518.1 XP_008759740.1 7 810
GenBank mRNA BC158723.1 - 3 291

View Full Product Details