Gene Transcripts

Gene Symbol
Kcnma1
Entrez Gene ID
83731
Gene Name
potassium channel, calcium activated large conductance subfamily M alpha, member 1
Gene Aliases
BKCa, KCNMA1b, KCNMA1c, Kcnma, Slo
Chromosome Location
Chr. 15: 344204 - 1048849 on Build Rnor_6.0
UniGene ID
Rn.30616
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031828.1 NP_114016.1 16, 18 2428
GenBank mRNA AF083341.1 - 16, 18 1864
AF135265.1 - 16, 18 2282
U40603.1 - 16, 18 2428

View Full Product Details