Gene Transcripts

Gene Symbol
Cyp4a8
Entrez Gene ID
266674
Gene Name
cytochrome P450, family 4, subfamily a, polypeptide 8
Gene Aliases
Cyp4a12
Chromosome Location
Chr. 5: 133978953 - 134008255 on Build Rnor_6.0
UniGene ID
Rn.10034
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031605.2 NP_113793.2 1 61
GenBank mRNA BC081771.1 - 1 61
M37828.1 - 1 51

View Full Product Details