Gene Transcripts

Gene Symbol
Epgn
Entrez Gene ID
289515
Gene Name
epithelial mitogen
Gene Aliases
RGD1560084
Chromosome Location
Chr. 14: 18627103 - 18635135 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_223280.6 XP_223280.3 4 561

View Full Product Details