Gene Transcripts

Gene Symbol
Hdhd1
Entrez Gene ID
291585
Gene Name
haloacid dehalogenase-like hydrolase domain containing 1
Gene Aliases
Fam16ax, Hdhd1a
Chromosome Location
Chr. 18: 45378365 - 45380778 on Build Rnor_6.0
UniGene ID
Rn.225791
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106146.1 NP_001099616.1 1 536
XM_006254718.2 XP_006254780.1 1 584

View Full Product Details