Gene Transcripts

Gene Symbol
Fam13a
Entrez Gene ID
362378
Gene Name
family with sequence similarity 13, member A
Gene Aliases
Fam13a1, RGD1309807
Chromosome Location
Chr. 4: 89182751 - 89281271 on Build Rnor_6.0
UniGene ID
Rn.47690
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001100862.1 NP_001094332.1 9 1050
XM_006236586.2 XP_006236648.1 8 1057
XM_008762964.1 XP_008761186.1 9 1084

View Full Product Details