Gene Transcripts

Gene Symbol
Olr7
Entrez Gene ID
405043
Gene Name
olfactory receptor 7
Gene Aliases
Chromosome Location
Chr. 1: 70484947 - 70485888 on Build Rnor_6.0
UniGene ID
Rn.142002
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001000768.1 NP_001000768.1 1 869

View Full Product Details