Gene Transcripts

Gene Symbol
Hp
Entrez Gene ID
24464
Gene Name
haptoglobin
Gene Aliases
Ba1-647
Chromosome Location
Chr. 19: 42096255 - 42100805 on Build Rnor_6.0
UniGene ID
Rn.10950
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012582.2 NP_036714.2 5 586
GenBank mRNA AF476963.1 - 5 617
AY325231.1 - 6 714
BC089816.1 - 5 586 7321960
FQ209575.1 - 3 491
FQ209684.1 - 5 638
FQ211144.1 - 5 640
FQ219122.1 - 5 638
FQ219413.1 - 2 396
FQ219443.1 - 5 638
FQ219547.1 - 5 609
FQ219598.1 - 5 608
K01933.1 - 2 350

View Full Product Details