Gene Transcripts

Gene Symbol
Gdf7
Entrez Gene ID
252833
Gene Name
growth differentiation factor 7
Gene Aliases
Chromosome Location
Chr. 6: 33496596 - 33507626 on Build Rnor_6.0
UniGene ID
Rn.92437
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001170350.1 NP_001163821.1 2 897
XM_006239878.2 XP_006239940.1 2 1002

View Full Product Details