Gene Transcripts

Gene Symbol
Nccrp1
Entrez Gene ID
292755
Gene Name
non-specific cytotoxic cell receptor protein 1 homolog (zebrafish)
Gene Aliases
RGD1305932
Chromosome Location
Chr. 1: 85213663 - 85216714 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001134506.2 NP_001127978.1 383
XM_008759218.1 XP_008757440.1 1, 2 383

View Full Product Details