Gene Transcripts

Gene Symbol
Sun5
Entrez Gene ID
296289
Gene Name
Sad1 and UNC84 domain containing 5
Gene Aliases
Spag4l
Chromosome Location
Chr. 3: 149336114 - 149356166 on Build Rnor_6.0
UniGene ID
Rn.218586
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106530.1 NP_001100000.1 9 754
XM_008762312.1 XP_008760534.1 11 925

View Full Product Details