Gene Transcripts

Gene Symbol
Ptprj
Entrez Gene ID
29645
Gene Name
protein tyrosine phosphatase, receptor type, J
Gene Aliases
DEP-1
Chromosome Location
Chr. 3: 79233519 - 79390956 on Build Rnor_6.0
UniGene ID
Rn.10278
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017269.2 NP_058965.2 3 403
GenBank mRNA U40790.1 - 3 333

View Full Product Details