Gene Transcripts

Gene Symbol
Crygb
Entrez Gene ID
301468
Gene Name
crystallin, gamma B
Gene Aliases
Cryg2, Len
Chromosome Location
Chr. 9: 71796204 - 71798265 on Build Rnor_6.0
UniGene ID
Rn.202960
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109875.1 NP_001103345.1 3 492

View Full Product Details