Gene Transcripts

Gene Symbol
Ttc28
Entrez Gene ID
304558
Gene Name
tetratricopeptide repeat domain 28
Gene Aliases
Chromosome Location
Chr. 12: 51385808 - 51840708 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006249584.2 XP_006249646.1 23 6876
XM_006249585.2 XP_006249647.1 22 7393

View Full Product Details