Gene Transcripts

Gene Symbol
Pvrl2
Entrez Gene ID
308417
Gene Name
poliovirus receptor-related 2
Gene Aliases
Chromosome Location
Chr. 1: 80631449 - 80666741 on Build Rnor_6.0
UniGene ID
Rn.203297
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012064.1 NP_001012064.1 8 1491
GenBank mRNA BC090075.1 - 8 1491

View Full Product Details