Gene Transcripts

Gene Symbol
Cnfn
Entrez Gene ID
365217
Gene Name
cornifelin
Gene Aliases
RGD1563577
Chromosome Location
Chr. 1: 82234032 - 82236414 on Build Rnor_6.0
UniGene ID
Rn.161807
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108909.1 NP_001102379.1 4 459
XM_008758960.1 XP_008757182.1 3 505

View Full Product Details