Gene Transcripts

Gene Symbol
Klk1c6
Entrez Gene ID
408242
Gene Name
kallikrein 1-related peptidase C6
Gene Aliases
Gk11, Gk6, Klk6, rGK-6
Chromosome Location
Chr. 1: 99947714 - 99964824 on Build Rnor_6.0
UniGene ID
Rn.141979
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001003977.1 NP_001003977.1 3 296
XM_006229110.2 XP_006229172.1 3 374

View Full Product Details