Gene Transcripts

Gene Symbol
Vom1r69
Entrez Gene ID
494251
Gene Name
vomeronasal 1 receptor 69
Gene Aliases
V1rc40
Chromosome Location
Chr. 4: 87455001 - 87455909 on Build Rnor_6.0
UniGene ID
Rn.129671
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001009531.1 NP_001009531.1 1 580
GenBank mRNA AY510330.1 - 1 580

View Full Product Details