Gene Transcripts

Gene Symbol
Vom1r95
Entrez Gene ID
494295
Gene Name
vomeronasal 1 receptor 95
Gene Aliases
V1ra7, Vmn1r52
Chromosome Location
Chr. 4: 121821837 - 121822781 on Build Rnor_6.0
UniGene ID
Rn.129646
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008950.1 NP_001008950.1 1 739
GenBank mRNA AY510341.1 - 1 739

View Full Product Details