Gene Transcripts

Gene Symbol
Arl5c
Entrez Gene ID
497990
Gene Name
ADP-ribosylation factor-like 5C
Gene Aliases
RGD1563128
Chromosome Location
Chr. 10: 85941174 - 85948131 on Build Rnor_6.0
UniGene ID
Rn.23998
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109046.1 NP_001102516.1 5 443
XM_006247396.2 XP_006247458.1 5 636
XM_006247397.2 XP_006247459.1 5 630

View Full Product Details