Gene Transcripts

Gene Symbol
Mfsd4
Entrez Gene ID
498228
Gene Name
major facilitator superfamily domain containing 4
Gene Aliases
RGD1566090
Chromosome Location
Chr. 13: 48807720 - 48848997 on Build Rnor_6.0
UniGene ID
Rn.213338
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109069.1 NP_001102539.1 7 1360
XM_006249791.2 XP_006249853.1 7 1355

View Full Product Details