Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001166303.1 | NP_001159775.1 | 3 | 641 | |
XM_006255877.2 | XP_006255939.1 | 3 | 1504 | ||
XM_008772699.1 | XP_008770921.1 | 3 | 722 | ||
GenBank mRNA | FQ213683.1 | - | 2 | 752 | |
FQ214505.1 | - | 3 | 701 |