Gene Transcripts

Gene Symbol
Noto
Entrez Gene ID
502857
Gene Name
notochord homeobox
Gene Aliases
RGD1562910
Chromosome Location
Chr. 4: 117206355 - 117210856 on Build Rnor_6.0
UniGene ID
Rn.228214
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001192014.1 NP_001178943.1 2 483

View Full Product Details