Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_172074.3 | NP_742071.1 | 2 | 732 | |
XM_006234473.2 | XP_006234535.1 | 3 | 344 | ||
XM_006234474.2 | XP_006234536.1 | 4 | 456 | ||
GenBank mRNA | AF150091.1 | - | 2 | 167 | |
BC091116.1 | - | 2 | 733 | 7366212 |