Gene Transcripts

Gene Symbol
Teddm1
Entrez Gene ID
664710
Gene Name
transmembrane epididymal protein 1
Gene Aliases
Chromosome Location
Chr. 13: 71322759 - 71324016 on Build Rnor_6.0
UniGene ID
Rn.195123
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001039915.2 NP_001035004.1 1 157
GenBank mRNA AJ515382.1 - 1 158

View Full Product Details