Gene Transcripts

Gene Symbol
Ttc32
Entrez Gene ID
684830
Gene Name
tetratricopeptide repeat domain 32
Gene Aliases
Chromosome Location
Chr. 6: 34154966 - 34161857 on Build Rnor_6.0
UniGene ID
Rn.216527
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_003750130.3 XP_003750178.1 2 300
GenBank mRNA FQ209446.1 - 1 24

View Full Product Details