Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SMAD9
Entrez Gene ID
694572
Gene Name
SMAD family member 9
Gene Aliases
EGK_09245
Chromosome Location
Chr.17: 15687619 - 15762045 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001257798.1 NP_001244727.1 - 906 136
GenBank mRNA JU320767.1 - - 906 136
JV715071.1 - - 364 136

View Full Product Details