Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FBXO31
Entrez Gene ID
695313
Gene Name
F-box protein 31
Gene Aliases
-
Chromosome Location
Chr.20: 74321888 - 74378352 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001258079.1 NP_001245008.1 - 1416 75
XM_015126750.2 XP_014982236.1 - 1541 75
XM_028840367.1 XP_028696200.1 - 1382 75
GenBank mRNA JU326513.1 - - 1249 75
JU477237.1 - - 1416 75

View Full Product Details