Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SERPIND1
Entrez Gene ID
696950
Gene Name
serpin family D member 1
Gene Aliases
EGK_02823
Chromosome Location
Chr.10: 31557250 - 31571352 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001265760.2 NP_001252689.1 - 1175 66
XM_015149746.2 XP_015005232.1 - 1434 66
GenBank mRNA JV504584.1 - - 853 66
JV600210.1 - - 1178 66

View Full Product Details