Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
THAP7
Entrez Gene ID
696208
Gene Name
THAP domain containing 7
Gene Aliases
-
Chromosome Location
Chr.10: 31797317 - 31799686 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001261590.1 NP_001248519.1 - 255 51
XM_015149745.2 XP_015005231.1 - 165 51
GenBank mRNA JU336514.1 - - 147 51
JU477325.1 - - 274 51
JV048319.1 - - 255 51
JV662341.1 - - 274 51

View Full Product Details