Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- NDE1
- Entrez Gene ID
-
717754
- Gene Name
- nudE neurodevelopment protein 1
- Gene Aliases
-
-
- Chromosome Location
-
Chr.20: 15223413 - 15307054 on Build Mmul_10
- Gene Symbol
- MYH11
- Entrez Gene ID
-
713882
- Gene Name
- myosin heavy chain 11
- Gene Aliases
-
-
- Chromosome Location
-
Chr.20: 15284858 - 15437191 on Build Mmul_10
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
JV652145.1 |
- |
- |
4811 |
65 |
View Full Product Details