Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LYAR
Entrez Gene ID
712499
Gene Name
Ly1 antibody reactive
Gene Aliases
-
Chromosome Location
Chr.5: 8651688 - 8674761 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001258141.1 NP_001245070.1 - 553 95
XM_015137914.2 XP_014993400.1 - 529 95
XM_015137915.2 XP_014993401.1 - 603 95
XM_015137916.2 XP_014993402.1 - 493 95
XM_015137918.2 XP_014993404.1 - 534 95
GenBank mRNA JU324146.1 - - 566 95
JU476323.1 - - 553 95
JV045791.1 - - 462 95

View Full Product Details