Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NADK2
Entrez Gene ID
704285
Gene Name
NAD kinase 2, mitochondrial
Gene Aliases
C6H5orf33, NADKD1
Chromosome Location
Chr.6: 35989627 - 36035149 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001092624.4 XP_001092624.2 - 525 108
XM_015139873.2 XP_014995359.1 - 526 108
XM_028849386.1 XP_028705219.1 - 210 108
XM_028849387.1 XP_028705220.1 - 322 108
XM_028849388.1 XP_028705221.1 - 210 108

View Full Product Details